Learn More
Restek Corporation Restek™ Split Liners for Thermo TRACE and Focus SSL
SDP
Catalog No. 06711276 Shop All Restek Corporation Products
Encompass_Preferred
Sleeve, Cup Splitter; 4mm I.D.; For Thermo TRACE and Focus SSL; Length: 105mm; 8.0mm O.D.
Catalog No. 06-711-276 Supplier Restek Corporation Supplier No. 20950
Sleeve, Cup Splitter; 4mm I.D.; For Thermo TRACE and Focus SSL; Length: 105mm; 8.0mm O.D.