Learn More
Restek Corporation Restek™ Split Liners for Thermo TRACE and Focus SSL
SDP
Supplier:  Restek Corporation 20937200.5
Encompass_Preferred
Sleeve, Split; 3mm I.D.; For Thermo TRACE and Focus SSL; Length: 105mm; 8.0mm O.D.; w/FS Wool; Pack of 5
Catalog No. 06-711-245