Learn More
Restek Corporation Restek™ Split Liners for Thermo TRACE and Focus SSL
SDP
Supplier:  Restek Corporation 20940202.5
Encompass_Preferred
Liner, split; Restek; Universal; ID: 5.0mm; OD: 8.0mm; Length: 105mm; With glass wool prepacked inlet liner; Pack of 5
Catalog No. 06-718-204