Learn More
Restek Corporation Restek™ Split Liners for Thermo TRACE and Focus SSL
Supplier: Restek Corporation 20939200.1
Sleeve, Split; 5mm I.D.; For Thermo TRACE and Focus SSL; Length: 105mm; 8.0mm O.D.; w/FS Wool