Learn More
Restek Corporation Restek™ Split Liners for Thermo TRACE and Focus SSL
Supplier: Restek Corporation 20937211.5
Sleeve, Split; 3mm I.D.; For Thermo TRACE and Focus SSL; Length: 105mm; 8.0mm O.D.; Base Deactivated w/Base Deactivated Wool; Pack of 5