Learn More
Restek Corporation Restek™ Split Liners for Thermo TRACE and Focus SSL
Supplier: Restek Corporation 20937202.5
Liner, split; Restek; Universal; ID: 3.0mm; OD: 8.0mm; Length: 105mm; With glass wool prepacked inlet liner; Pack of 5