Learn More
Restek Corporation Restek™ Split Liners for Thermo TRACE and Focus SSL
SDP
Supplier:  Restek Corporation 20941211.25
Encompass_Preferred
Sleeve, Split; 5mm I.D.; For Thermo TRACE and Focus SSL; Length: 105mm; 8.0mm O.D.; Base Deactivated w/Base Deactivated Wool; Pack of 25
Catalog No. 06-711-257