Learn More
Restek Corporation Restek Splitless Liners for Agilent/Finnigan: 2mm Splitless
SDP
Supplier:  Restek Corporation 20714214.25
Encompass_Preferred
Sleeve, Splitless; 2mm I.D.; For Agilent/Finnegan GCs; Length: 78.5mm; 6.5mm O.D.; Siltek Deactivated; Pack of 25
Catalog No. 06-717-838