Learn More
Restek Corporation Restek Splitless Liners for Agilent/Finnigan GCs: Recessed Gooseneck
Supplier: Restek Corporation 20984209.5
Sleeve, Recess Taper Gooseneck; 4mm I.D.; Length: 78.5mm; 6.5mm O.D.; For Agilent/Finnegan GCs; w/CarboFrit; Pack of 5