Learn More
Restek Corporation Restek Splitless Liners for Agilent/Finnigan: Gooseneck Splitless
Supplier: Restek Corporation 20800216.25
Sleeve, Splitless Gooseneck; 4mm I.D.; Length: 78.5mm; 6.5mm O.D.; For Agilent/Finnegan GCs; Siltek Deactivated w/Carbofrit; Pack of 25