Learn More
Restek Corporation Restek Splitless Liners for Varian 1075/1077 GCs
SDP
Catalog No. 06711202 Shop All Restek Corporation Products
Encompass_Preferred
Sleeve, Splitless; 4mm I.D.; For Varian 1075/1077 GCs; Length: 74mm; 6.3mm O.D.
Catalog No. 06-711-202 Supplier Restek Corporation Supplier No. 20904
Sleeve, Splitless; 4mm I.D.; For Varian 1075/1077 GCs; Length: 74mm; 6.3mm O.D.