Learn More
Restek Corporation Restek Splitless Liners for Varian 1075/1077 GCs
SDP
Supplier:  Restek Corporation 20905214.5
Encompass_Preferred
Sleeve, Splitless; 4mm I.D.; For Varian 1075/1077 GCs; Length: 74mm; 6.3mm O.D.; Siltek Deactivated; Pack of 5
Catalog No. 06-711-211