Learn More
Restek™ Sulfinert Sample Loops
SDP
Ideal for samples containing low concentrations of sulfur or other active compounds. Sulfinert treatment of Restek Gas Sample Loops eliminate active sites in the valve or loop for better recovery of active compounds.
$406.50 - $421.50
Specifications
Type Gas Sample Loop
Material Sulfinert treated
View More Specs
Products 11
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
06-712-059
SDP
View Documents
Restek™
22842
20 μL
Each for $406.50
Only null left
Add to Cart
06-712-058
SDP
View Documents
Restek™
22841
10 μL
Each for $406.50
Only null left
Add to Cart
06-712-060
SDP
View Documents
Restek™
22843
25 μL
Each for $421.50
Only null left
Add to Cart
06-712-057
SDP
View Documents
Restek™
22840
5 μL
Each for $406.50
Only null left
Edge
Add to Cart
06-712-062
SDP
View Documents
Restek™
22845
100 μL
Each for $421.50
Only null left
Add to Cart
06-712-061
SDP
View Documents
Restek™
22844
50 μL
Each for $421.50
Only null left
Edge
Add to Cart
06-712-063
SDP
View Documents
Restek™
22846
250 μL
Each for $421.50
Only null left
Add to Cart
06-712-064
SDP
View Documents
Restek™
22847
500 μL
Each for $406.50
Only null left
Add to Cart
06-712-065
SDP
View Documents
Restek™
22848
1 mL
Each for $406.50
Only null left
Edge
Add to Cart
06-712-066
SDP
View Documents
Restek™
22849
2 mL
Each for $421.50
Only null left
Add to Cart
06-712-067
SDP
View Documents
Restek™
22850
5 mL
Each for $406.50
Only null left
Add to Cart
Description
Description
  • Sample loop sizes from 5μL to 5mL
  • 0.0625 in. fittings
  • 1mm I.D.
  • For “W Type” Valves
Specifications
Specifications
Gas Sample Loop
Sulfinert treated
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.