Learn More
Restek™ Super-Clean Gas Trapping System for LC/MS
SDP
Catalog No. p-3218366 Shop All Restek Corporation Products
Change view
Click to view available options
Type:
Gas Trapping System
Hydrocarbon Filter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-711-863 Hydrocarbon Filter
06-711-864 Gas Trapping System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-711-863 Supplier Restek™ Supplier No. 22061
Only null left
Add to Cart
Add to Cart
Quick-change cartridge system removing hydrocarbons efficiently from nitrogen
  • Changing filters is quick and easy
  • Up to 20L of hydrocarbon-free nitrogen per minute
  • Filters connected in parallel to handle high flows of LC/MS
Specifications
Type Hydrocarbon Filter
For Use With (Equipment) Super-Clean Gas Trapping System