Learn More
Restek™ Syringe Adapter Kit
SDP
For Single-Stage VOC Regulator
Supplier:  Restek™ 24613
 View more versions of this product
Catalog No. 06-712-389
Includes:
One nickel-plated brass 1/4 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, One stainless steel, 1/4 in. NPT x 1/8 in. compression fitting with septum (can be used with any syringe needle).
Description
Description
  • Use to withdraw sample from a high-pressure cylinder after pressure reduction through the high-purity VOC single-stage regulator
Specifications
Specifications
Chaney Adapter
Use to withdraw sample from a high pressure cylinder after pressure reduction through the high purity VOC single stage regulator
700/1700/1000/5 to 10μL syringes
One Nickel-plated Brass 0.25 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, one Stainless Steel, 0.25 in. NPT x 0.125 in. compression fitting with septum (can be used with any syringe needle)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.