Learn More
Restek™ Tenax Packings
SDP
Catalog No. 06713069 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Particle Size:
20/35 Mesh
35/60 Mesh
60/80 Mesh
80/100 Mesh
Material:
Tenax-TA
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Material
06-713-069 35/60 Mesh Tenax-TA
06-712-954 80/100 Mesh Tenax-TA
06-712-953 60/80 Mesh Tenax-TA
06-713-068 20/35 Mesh Tenax-TA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-713-069 Supplier Restek™ Supplier No. 25701
Porous polymer resin packing for GC columns.
  • Tenax Packing Materials
  • Minimum order: 10 grams
Specifications
Particle Size 35/60 Mesh
Product Type Packing Material
Material Tenax-TA