Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ TO-14A Aromatics Mix
SDP

Catalog No. 06802388 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Concentration:
1 ppmv
100 ppbv
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
06-802-388 100 ppbv
06-802-377 1 ppmv
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-802-388 Supplier Restek™ Supplier No. 34423
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Certificate of Analysis and unique serial number

Disposable and does not require rental or demurrage fees

Air-monitoring ga calibration standards are provided by Spectra/Linde

  • Minimum 12-month stability on all cylinders
  • Aluminum construction
  • Size: 8 x 24cm
  • Volume/Pressure: 104L of gas at 1800 psi
  • CGA-180 outlet fitting
  • Weight: 1.5 lb. (0.7kg)

Compounds

  • Benzene
  • Chlorobenzene
  • m-Dichlorobenzene
  • o-Dichlorobenzene
  • p-Dichlorobenzene
  • Ethyl benzene
  • Styrene
  • Toluene
  • 1,2,4-Trichlorobenzene
  • 1,2,4-Trimethylbenzene
  • 1,3,5-Trimethylbenzene
  • m-Xylene
  • o-Xylene
  • p-Xylene

Order Info

Gas standards are subject to hazardous materials shipping fees by most freight carriers. All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements. Drop-shipped to provide the freshest standard possible.

Specifications

Components Benzene, Chlorobenzene, m-Dichlorobenzene, o-Dichlorobenzene, p-Dichlorobenzene, Ethyl Benzene, Styrene, Toluene, 1,2,4-Trichlorobenzene, 1,2,4-Trimethylbenzene, 1,3,5-Trimethylbenzene, m-Xylene, o-Xylene, p-Xylene in Nitrogen
Concentration 100 ppbv
Volume (Metric) Gas 104 L
Pressure 1800 psi
Weight (English) 1.5 lb.
Volume (English) Gas 27.47 gal.
Weight (Metric) 0.7 kg
Type TO-14A Aromatics Mix
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.