Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ TO-14A Internal Standard/Tuning Mix

Cylinders are disposable and do not require rental or demurrage fees
$1925.00 - $2269.00
Specifications
Components | Bromochloromethane, 4-Bromofluorobenzene, Chlorobenzene-d5, 1,4-Difluorobenzene in Nitrogen |
---|---|
Volume (Metric) Gas | 104 L |
Pressure | 1800 psi |
Weight (English) | 1.5 lb. |
Volume (English) Gas | 27.47 gal. |
Includes: Certificate of Analysis and unique serial number
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
06-802-379
![]() |
Restek™
34408 |
1 ppm |
Each for $1,925.00
|
|
|||||
06-802-390
![]() |
Restek™
34425 |
100 particles/mL |
Each for $2,269.00
|
|
|||||
Description
Calibration standards are provided by Spectra/Linde.
- Meets lab requirements for two separate sources of calibration standards
- Minimum 12-month stability on all cylinders
- Aluminum construction
- Size: 8 × 24cm
- Volume/Pressure: 104L of gas at 1800 psi
- CGA-180 outlet fitting
- Weight: 0.7kg (1.5 lb.)
Compounds
- Bromochloromethane
- 1-Bromo-4-fluorobenzene (4-Bromofluorobenzene)
- Chlorobenzene-d5
- 1,4-Difluorobenzene
Order Info
Gas standards are subject to hazardous materials shipping fees by most freight carriers. All calibration gas standards are nonreturnable due to DOT hazardous shipping requirements.
Specifications
Bromochloromethane, 4-Bromofluorobenzene, Chlorobenzene-d5, 1,4-Difluorobenzene in Nitrogen | |
1800 psi | |
27.47 gal. | |
TO-14A Internal Standard/Tuning Mix |
104 L | |
1.5 lb. | |
0.7 kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title