Learn More
Restek™ Tubing Cutter
SDP
Catalog No. 06700003 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
To cut 4 to 28mm OD tubing.
Catalog No. For Use With (Application)
06-700-003 To cut 4 to 28mm OD tubing.
1 options
Catalog No. 06-700-003 Supplier Restek™ Supplier No. 22356
Designed to cut 4 to 28mm OD tubing.
  • Easily cuts stainless steel, Monel alloy, and other hard-temper materials
  • Unique design makes clean, right-angle cuts and eliminates spiraling
  • Integral reamer folds away for safety and convenience
  • Replacement cutter wheels also available
Specifications
For Use With (Application) To cut 4 to 28mm OD tubing.
Product Type Tubing Cutter