Description
- Equivalent to XAD-2 Resin
- For adsorbing semivolatiles in air
- Cleaned and GC tested
- Quantity: 100g
Specifications
Specifications
| Quantity | 100 g |
| Product Type | Ultra Clean Resin |