Learn More
Restek Corporation Restek Undeactivated, Untreated Fused Silica Tubing
SDP
Catalog No. 06717553 Shop All Restek Corporation Products
Encompass_Preferred
Tubing, Undeactivated, Untreated Fused Silica Tubing; Restek; Coiled in 7 in. diameters, banded in 5m sections, mounted in a Restek cage; 60m length; 0.25mm I.D. x 0.37mm O.D. (plus or minus 0.04mm); Pack of 6
Catalog No. 06-717-553 Supplier Restek Corporation Supplier No. 10052600
Tubing, Undeactivated, Untreated Fused Silica Tubing; Restek; Coiled in 7 in. diameters, banded in 5m sections, mounted in a Restek cage; 60m length; 0.25mm I.D. x 0.37mm O.D. (plus or minus 0.04mm); Pack of 6