Learn More
Cayman Chemical trans-Resveratrol, 501-36-0, 500 mg
Supplier: Cayman Chemical 70675500
A polyphenolic phytoalexin found in a variety of plants; has anti-inflammatory, antioxidant, and anticancer activities; inhibits the cyclooxygenase and hydroperoxidase activities of COX-1 (EC50s = 15 and 3.7 μM, respectively), but not COX-2 (EC50s = >100 μM and 85 μM, respectively); inhibits carrageenan-induced paw edema in mice at 3 and 8 mg/kg; inhibits free radical formation in HL-60 human promyelocytic leukemia cells induced by TPA (EC50 = 27 μM); reduces both the incidence and number of tumors in a two-stage mouse model of skin cancer induced by TPA and DMBA at 1-25 μmol; activates SIRT1 by 8-fold in vitro at 200 μM; inhibits ERK1, JNK1, Src, PKCα, aromatase/CYP19, and DNA polymerases α and δ (IC50s = 37, 50, 20, in vitro and/or ex vivo; prolongs lifespan in model organisms including C. elegans, D. melanogaster, and mice
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.