Learn More
STEMCELL Technologies Resveratrol, Size: 500 mg
SDP
Supplier:  STEMCELL Technologies 72864
Encompass_Preferred
Resveratrol is a potent phenolic antioxidant found in grapes and red wine that also has antiproliferative and anti-inflammatory activity (Rotondo et al.). Resveratrol is also a selective inhibitor of cyclooxygenase-1 (COX-1; Jang) with an ED₅₀ value of 15 μM, respectively.MAINTENANCE AND SELF-RENEWAL· Inhibits differentiation of pig preadipocytes into adipocytes (Bai et al.).· Promotes self-renewal of human mesenchymal stem cells (Peltz et al.).DIFFERENTIATION· Induces osteogenic differentiation in human bone marrow-derived mesenchymal stem cell cultures (Boissy et al.; Dai et al.; Peltz et al.).· Induces osteoblast differentiation while blocking adipocyte development in mouse mesenchymal stem cells and cultured primary rat bone marrow cells (Bäckesjö et al.).
Catalog No. 50-197-5992
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.