Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resveratrol from Synthetic, ≥98%, MP Biomedicals™

Catalog No. ICN19605225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19605225 25 mg
ICN19605280 0.1 mg
ICN19605205 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19605225 Supplier MP Biomedicals, Inc Supplier No. 0219605225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 501-36-0
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00133799
InChI Key LUKBXSAWLPMMSZ-OWOJBTEDSA-N
Synonym resveratrol, trans-resveratrol, 3,4',5-trihydroxystilbene, 3,5,4'-trihydroxystilbene, e-resveratrol, 3,4',5-stilbenetriol, e-5-4-hydroxystyryl benzene-1,3-diol, resvida, 3,4',5-trihydroxy-trans-stilbene, 5-e-2-4-hydroxyphenyl ethenyl benzene-1,3-diol
PubChem CID 445154
ChEBI CHEBI:45713
IUPAC Name 5-[(E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
SMILES C1=CC(=CC=C1C=CC2=CC(=CC(=C2)O)O)O

Specifications

Color White
Assay Percent Range ≥98%
Quantity 25 mg
Formula Weight 228.25
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Resveratrol, from Synthetic
Recommended Storage Store at -20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.