accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHHO(E)

CAS RN 501-36-0

IUPAC Name: 5-[(1E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
Synonyms: resveratrol trans-resveratrol 3,4',5-trihydroxystilbene 3,4',5-stilbenetriol resveratrol (substance) RESV resveratrol-3-sulfate (E)-resveratrol 3,4',5-trihydroxy-stilbene
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: LUKBXSAWLPMMSZ-OWOJBTEDSA-N
SMILES: OC1=CC=C(\C=C\C2=CC(O)=CC(O)=C2)C=C1
Percent Purity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (7)
Physical Form 
  • (3)
  • (5)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (7)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)