Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Ret Oncoprotein Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1120R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1120R1 1 mL Prediluted
MS1120S0 100 μL Concentrated
MS1120S 1 mL Concentrated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1120R1 Supplier Epredia™ Supplier No. MS1120R1

Provide accurate, reproducible results with the Epredia™ Ret Oncoprotein Ab-1, Mouse Monoclonal Antibody.

The ret proto-oncogene encodes a cell surface glycoprotein belonging to a member of the receptor tyrosine kinase family and is located on chromosome 10q11.2. Three main 3' splice isoforms have been characterized from papillary thyroid carcinomas, themselves originating from thyroid epithelial cells. Ret expression is observed in several regions of the central nervous system; in the developing cranial nerve ganglia and a subset of cells within dorsal root ganglia, in motor neurons in the spinal cord and hindbrain, in neuroretina and the growing tips of the renal collecting ducts in developing kidney.

Host Species: Mouse

Clone: 3F8

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: Recombinant protein encoding the extreme C-terminal cytoplasmic region of the human ret oncoprotein

Positive Control: Enteric ganglion cells, Small intestine

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO


Specifications

Antigen Ret Oncoprotein Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3F8
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the extreme C-terminal cytoplasmic region of the human ret oncoprotein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.