Learn More
Synthware™ Replacement Plug Retention Kit
GREENER_CHOICESDP
Catalog No. 31502038 Shop All Kemtech Products
Change view
Click to view available options
Plug Bore:
12/30 mm
16.2/56 mm
17.35/50 mm
17/40 mm
20/44 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Bore
31-502-038 17.35/50 mm
31-502-036 12/30 mm
31-502-037 17/40 mm
31-502-039 16.2/56 mm
31-502-040 20/44 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31-502-038 Supplier Synthware™ Supplier No. S691750
Add to Cart
Add to Cart
For use with stopcocks
Specifications
Type Replacement Stainless Steel Retention Kit
Plug Bore 17.35/50 mm
Material Stainless Steel
For Use With (Application) Organic Synthesis
For Use With (Equipment) Glass Stopcock