Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Retinol Binding Protein ELISA Kit

Catalog No. EIARBP
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIARBP 96 Tests
EIARBPX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIARBP Supplier Invitrogen™ Supplier No. EIARBP
Add to Cart
Add to Cart

ELISA

The Retinol Binding Protein Competitive ELISA quantitates retinol binding protein (RBP) in serum, plasma, urine, saliva, dried fecal extracts or cell culture medium.∣Principle of the method: The Retinol Binding Protein Competitive ELISA research-use-only kit is designed to quantitatively measure RBP independent of species. An RBP standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-sheep IgG antibodies. Anti-sheep RBP antibodies and RBP-peroxidase conjugate is added to the wells. The RBP-peroxidase conjugate and any RBP in the sample will compete to bind to the anti-sheep RBP antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound RBP-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of RBP present in the sample.∣Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09455
Assay Range 7.8-1000 ng/mL
Assay Sensitivity 1.36 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine, Whole Blood
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias C CRABP-I; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRABP-I; CRBP; CRBP1; Crbpi; CRBP-I; LOW QUALITY PROTEIN: retinol-binding protein 1; mCRBPI; RBP; RBP1; Rbp-1; RBPC; retinol; retinol binding; Retinol Binding Protein; retinol binding protein 1; retinol binding protein 1, cellular; retinol-binding protein 1; retinol-binding protein 1, cellularC CRABP-I; cellular retinol binding protein 1; Cellular retinol-binding protein; Cellular retinol-binding protein I; CRABP-I; CRBP; CRBP1; Crbpi; CRBP-I; LOW QUALITY PROTEIN: retinol-binding protein 1; mCRBPI; RBP; RBP1; Rbp-1; RBPC; retinol; retinol binding; Retinol Binding Protein; retinol binding protein 1; retinol binding protein 1, cellular; retinol-binding protein 1; retinol-binding protein 1, cellular;
Gene ID (Entrez) 5947
Gene Symbol Rbp1
Interassay CV 12%
Intraassay CV 5%
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Shipping Condition Wet or Dry Ice
Storage Requirements 2-8°C
Target RBP1
Test Time 45 min
Total Assay Time 1 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.