Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 54884 |
| Product Line | Novus |
| Purification Method | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | Retinol Saturase Recombinant Protein Antigen |
| Content And Storage | Store at −20C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Alias | All-trans-13,14-dihydroretinol saturase, all-trans-retinol 13,14-reductase, EC 1.3.99.23, FLJ20296, retinol saturase (all-trans-retinol 13,14-reductase), RetSat |
| Gene Symbol | RETSAT |
| Label Type | Unlabeled |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
