Learn More
Swann Morton Retracting Guard Safety Scalpel

Description
- Stainless Steel
- Designed for tissue separation, puncturing or cutting procedures
- Easy one-handed activation
- OSHA compliant
- Covered blades reduces risk of accidental contact
Specifications
Specifications
| Product Type | Safety Scalpel |
| CAS | 67-56-1 |
| Size | # 10 |
| Disposable | Single-use |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| Graduated | Yes |
| Linear Formula | C11H17NO · HCl |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol |
| Concentration | 1.0 mg/mL in methanol (as free base) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.