Learn More
RETSCH Comfort Clamping Device for AS 200 Sieve Shakers
Catalog No. 0841769 Shop All RETSCH Inc. Products
Change view
Click to view available options
For Use With (Equipment):
AS 200 sieve shaker
Catalog No. For Use With (Equipment)
08-417-69 AS 200 sieve shaker
1 options
Catalog No. 08-417-69 Supplier RETSCH Supplier No. 326620001
Add to Cart
Add to Cart
Provides reliable, quick and convenient clamping of sieves in dry operation.
  • View cover, smooth rods, and quick-action clamps
  • Cover stays on rods
  • Removable sieve stack
Specifications
Holds Test Sieves, 200 to 203 mm dia.
Product Type Comfort Clamping Device
For Use With (Application) Reliable, quick, convenient sieve clamping in dry operation
For Use With (Equipment) AS 200 sieve shaker