Learn More
RETSCH AS Series Sieve Shaker Accessory, Standard Clamping Device
Catalog No. 0841768 Shop All RETSCH Inc. Products
Change view
Click to view available options
For Use With (Equipment):
AS Series
Catalog No. For Use With (Equipment)
08-417-68 AS Series
1 options
Catalog No. 08-417-68 Supplier RETSCH Supplier No. 326620002
Add to Cart
Add to Cart
Provide reliable, quick and convenient clamping of sieves in dry operation
Provide reliable, quick and convenient clamping of sieves in dry operation.
  • Threaded rods and clamping nuts
  • View cover
Specifications
Product Type Clamp
For Use With (Application) Reliable, quick, convenient clamping of sieves in dry operation; For Sieves: 4-8mm
For Use With (Equipment) AS Series