Learn More
RETSCH ZM 200 Ultra Centrifugal Mill, Push-Fit Rotors
Catalog No. 084165 Shop All RETSCH Inc. Products
Change view
Click to view available options
Type:
12 Tooth SS - Medium Coarse Materials
6 Tooth Rotor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
08-416-5 6 Tooth Rotor
08-416-4 12 Tooth SS - Medium Coarse Materials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-416-5 Supplier RETSCH Supplier No. 026080040
Add to Cart
Add to Cart
Request a Quote
For fine grinding of soft to medium-hard and fibrous materials.
  • Accessory for use with ZM 200 Ultra Centrifugal Mill
  • Wear resistant coating
Specifications
Material Stainless Steel
Type 6 Tooth Rotor
Max. Speed 6,000 to 18,000 rpm
Max. Tube Diameter 9.9 cm
For Use With (Equipment) Centrifugal Mill