Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desco™ Reusable Attention Label
SDP

Catalog No. 18688004
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-688-004 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-688-004 Supplier Desco™ Supplier No. 06730
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Attention Label with non-binding adhesive, which allows the label to be removed easily.

  • ESD Susceptibility Symbol (RS-471) identifies the contents of the packaging as ESD susceptible
  • Reusable
  • Non-binding adhesive will allow the label to be removed and leaving little or no residue
  • Core: 3 in.

Specifications

Autoclavable Not Autoclavable
Adhesive Type Removable
Color Yellow
Material Paper
Sterility Non-sterile
Display Text Attention: Observe Precautions for Handling Electrostatic Sensitive Devices
For Use With (Application) Packages that are ESD susceptible
Latex Free Yes
Quantity 500/Roll
Dimensions (L x W) 2 x 2 in. (50.5 x 50.8 mm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.