Learn More
Thermo Scientific™ Reusable Calibrated Inoculating Loops
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Reusable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
- Twisted nichrome made
- With or without insulated handle
- Available in different color codes
Specifications
Specifications
| Material | Nichrome |
| Size | 2 mm |
| Color | White |
| Product Type | Inoculating Loop |
| Quantity | Each |
| Handle Type | White Insulated Handle |