Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CritiCore Protective Wear Integrity 1800 Cleanroom Coveralls

Catalog No. 18111271
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-271 Small
18-111-269 2X-Small
18-111-270 X-Small
18-111-272 Medium
18-111-273 Large
18-111-274 X-Large
18-111-275 2X-Large
18-111-276 3X-Large
18-111-277 4X-Large
18-111-278 5X-Large
18-111-279 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 18-111-271 Supplier CritiCore Protective Wear Supplier No. 13C23065N18001SWHT
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reusable coveralls with ESD qualities provide maximal fluid resistance and particle filtration for a variety of cleanroom applications including pharmaceutical, biotechnology, and medical device manufacturing.

  • Fluid and bacteria resistant Integrity 1800™ high performance carbon striped fabric
  • Mandarin collar with two snaps at top
  • Full-length, lint-limiting front zipper
  • Three male snaps on the back of the leg for boot attachment
  • Laser-cut, pre-serged seams
  • Silicon-free
  • ESD carbon stripes provide electrostatic dissipation

Specifications

Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Polyester, Carbon
Color White
Disposable Reusable
Ankle Style Snap
Cuff Style Knit
Neck Style Collared
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Anti-static, Liquid
Description Integrity™ 1800
Latex Free Yes
No. of Pockets None
Sleeve Style Raglan
For Use With (Application) Pharmaceutical, Biotechnology, Medical Device Manufacturing
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.