Learn More
CritiCore Protective Wear Reusable Double-Layer Integrity 1800 Tricot Face Mask
Catalog No. 18111379
Change view
Click to view available options
Color: White
Catalog No.
18-111-379 Color: White
1 options
This item is not returnable. View return policy
Catalog No. 18-111-379 Supplier CritiCore Protective Wear Supplier No. PD 6676
This item is not returnable. View return policy
General-purpose use mask that is domestically manufactured. Mask is not suitable for virologic or bacterial protection.
  • Washable
  • 100% filament polyester
  • One layer of Integrity 1800 cleanroom fabric
  • One layer of Tricot surgical fabric
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Material Tricot Fabric
Model PD 6676