Learn More
World Precision Instrument Reusable SurgiClose Skin Stapler, High Temp Autoclavable
SDP
Supplier:  World Precision Instrument 504564
Encompass_Preferred
The SurgiClose™ REP is a reusable device which allows for reloading of sterile, multi-feed staples and can be re-sterilized (by autoclave, gas and chemical means) for approximately 100 procedures.
Catalog No. 50-253-9338
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.