Learn More
PIP™ Reusable TPR Corded Ear Plugs - NRR 25
Catalog No. 17489659 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Noise Reduction Rating (NRR):
25
Catalog No. Noise Reduction Rating (NRR)
17-489-659 25
1 options
Catalog No. 17-489-659 Supplier PIP™ Supplier No. 267HPR320C
  • $138.00
    $79.65 / Pack of 100
    Save $58.35
    42% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Reusable TPR corded ear plugs - NRR 25.
  • Pre-formed plug shape with no rolling required makes insertion simple
  • Self-sealing flanges give all day comfort
  • Rigid stem makes insertion easy and reduces the risk of contamination
  • Corded versions use a non-resonating textile cord for comfort and easy storage
  • Cord can be detached for added safety when working around machines, or when the TPR plug is to be cleaned using soap and water
  • Can be reused several times, making them very cost efficient
  • Moderate attenuation allows for safe usage that blocks out hazardous noise yet allows warning signals and voices to be clearly understood
Specifications
Size Universal
Noise Reduction Rating (NRR) 25
Corded Corded (detachable)
Plug Shape Flanged
Product Type Reusable Corded Ear Plugs
Disposable Reusable
Metal Detectable No
Dielectric Yes