Description
- Pre-formed plug shape with no rolling required makes insertion simple
- Self-sealing flanges give all day comfort
- Rigid stem makes insertion easy and reduces the risk of contamination
- Corded versions use a non-resonating textile cord for comfort and easy storage
- Cord can be detached for added safety when working around machines, or when the TPR plug is to be cleaned using soap and water
- Can be reused several times, making them very cost efficient
- Moderate attenuation allows for safe usage that blocks out hazardous noise yet allows warning signals and voices to be clearly understood
Specifications
Specifications
| Size | Universal |
| Noise Reduction Rating (NRR) | 27 |
| Corded | Corded (detachable) |
| Plug Shape | Flanged |
| Product Type | Reusable Corded Ear Plugs |
| Disposable | Reusable |
| Metal Detectable | No |
| Dielectric | Yes |