Learn More
Description
- The corrosion-resistant stainless construction includes rolled top rims, smooth edges and radiused corners for easy, thorough cleaning.
- These beakers do not contain graduation markings - designed to be filled to the brim to realize full vessel capacity.
- Useful for storing, mixing, and heating solutions in laboratory and production settings when durability and easy sterilization is desired.
- Available in sizes from 100 mL to 5000 mL. Sold individually.
- REUZ™ products from United Scientific promote sustainability in all laboratory environments.
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| Material | Stainless Steel (304) |
| Graduated | Yes |
| Style | Griffin, Low Form |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


