Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ REUZ™ Stainless Steel Scoop
SDPSCI_ED

Catalog No. S139213 Shop All United Scientific Supplies, Inc Products
Click to view available options
Capacity (Metric):
100 mL
200 mL
250 mL
350 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

REUZ™ products from United Scientific promote sustainability in all laboratory environments.

  • Rounded bowls of the scoops are completely seamless for easy cleaning.
  • The corrosion-resistant stainless construction includes large handles and smooth, large radius interior surfaces for easy cleaning and protection against bacteria deposits.
  • Available in sizes from 100 mL to 500 mL.
  • Sold individually.
  • REUZ™ products from United Scientific promote sustainability in all laboratory environments.

Specifications

Product Type Scoop
Material Stainless Steel
Autoclavable Autoclavable
Capacity (Metric) 350 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.