Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Black Stallion™ Tool Handz™ Plus Original Mechanics Gloves

Catalog No. 19840301
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-840-301 Medium
19-840-302 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-840-301 Supplier Black Stallion™ Supplier No. 99PLUSBLKM
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves are constructed of lightweight and durable synthetic leather with a reinforced palm and fingertips.

  • Titan 2 Synthetic Leather Palm
  • Reinforced Contact Zones
  • Spandex Back
  • Strap Cuff

Specifications

Back Material Spandex
Cuff Material Elastic
Coating Material Uncoated
Color Black
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size Medium
For Use With (Application) Automotive Assembly, Construction, and Industrial Environments
Series 99PLUS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.