Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Reversan
GREENER_CHOICESDP

Catalog No. 372210R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-221-0R 10 mg
37-225-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-221-0R Supplier Tocris Bioscience™ Supplier No. 3722/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective MRP1 and P-gp inhibitor

Reversan is a selective inhibitor of multidrug resistance-associated protein 1 (MRP1) and P-glycoprotein (P-gp). Increases the sensitivity of MRP1-overexpressing tumor cells (MCF7/VP) to doxorubicin (Cat. No. 2252), vincristine (Cat. No. 1257) and etoposide (Cat. No. 1226) by 3.8-, 14.6- and 11.6-fold respectively. Increases the efficacy of vincristine and etoposide in murine models of neuroblastoma in vivo. Does not sensitize MRP2, MRP3, MRP4 or MRP5 transporters to known substrates.

Specifications

Quantity 10 mg
Product Type Reversan
Purity 0.95
Molecular Weight (g/mol) 441.52
Inhibitors Reversan

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.