Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio RevIT AAV Enhancer, 75 mL, for 50 L of Cell Culture

Catalog No. MIR8080 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR8080 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MIR8080 Supplier Mirus Bio Supplier No. MIR 8080
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Paired with any transfection reagent, the RevIT AAV Enhancer delivers higher titers for recombinant adeno-associated virus production

  • This novel enhancer is easily integrated into existing workflows, and it produces 2 to 4x higher titers across a range of serotypes, cellular growth media formulations, and transfection platforms.
  • RevIT™ AAV Enhancer can further drive down upstream manufacturing costs by permitting the use of lower levels of plasmid DNA while maintaining high-quality titers.
  • Continual improvements in both upstream and downstream AAV manufacturing processes will allow the next generation of gene therapies to reach a broader audience.

Specifications

Cell Type 293-derived Cells
Format Bottle
Promoter None (Promoterless)
Transfection Technique Lipid-based Transfection
For Use With (Application) AAV Production
Quantity 75 mL
Sterility Sterile
Volume (Metric) 75 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.