Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rex1 Recombinant Superclonal™ Antibody (14HCLC)

Catalog No. PI710190 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI710190 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI710190 Supplier Invitrogen™ Supplier No. 710190
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Superclonal Antibody

Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

Involved in the reprogramming of X-chromosome inactivation during the acquisition of pluripotency. Required for efficient elongation of TSIX, a non-coding RNA antisense to XIST. Binds DXPas34 enhancer within the TSIX promoter. Involved in ES cell self-renewal. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rex1
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Clone 14HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene ZFP42
Gene Accession No. Q96MM3
Gene Alias 1700021P10Rik; 2610511M11Rik; AW050347; ELOABP1; EloA-BP1; elongin A binding protein 1; elongin A-binding protein 1; elongin-A-binding protein 1; hREX-1; KIAA1138; mREX-1; reduced expression protein 1; REX1; Rex-1; REX1 transcription factor; REX1, RNA exonuclease 1 homolog; REX1, RNA exonuclease 1 homolog (S. cerevisiae); REXO1; RNA exonuclease 1 homolog; Tceb3bp1; transcription elongation factor B polypeptide 3 binding protein 1; transcription elongation factor B polypeptide 3-binding protein 1; Zfp42; Zfp-42; ZFP42 zinc finger protein; Zfp971; Zinc finger protein 42; zinc finger protein 42 homolog; Zinc finger protein 754; zinc finger protein 971; ZNF754
Gene Symbols ZFP42
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-140 of human REX1 transcription factor.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100361947, 100856057, 132625
Target Species Human, Rat, Canine, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.