Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desco™ Reztore Surface and Mat Cleaner, 1 Quart Spary
SDP

Catalog No. 18688044
Encompass
Encompass_Preferred
Change view
Click to view available options
Includes:
1 qt. trigger spray bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
18-688-044 1 qt. trigger spray bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-688-044 Supplier Desco™ Supplier No. 10446
May include imposed supplier surcharges.
Add to Cart
Add to Cart

ESD Surface and Mat Cleaner is used to clean and maintain ESD mats and other worksurfaces.

  • Removes dust, grease, grime, fingerprints, solder flux and other contaminants from ESD mats and other surfaces
  • Alcohol-free formula
  • Excessive use of alcohol-based cleaners may dry-out mats (vinyl or rubber) and degrade performance
  • Non-hazardous, non-flammable green chemistry formula
  • No special handling requirements for shipping and storage; may be used in normal workplace environments
  • No silicone or insulative residues
  • Cleaners with silicone leave an insulative layer preventing conductive or dissipative mats from functioning properly
  • Reztore™ Refill System Available
  • Reuse bottles and save cost by using the Desco 10448 Reztore™ 2.5 Gallon Bag-in-Box
  • Lead-free RoHS compliant

Specifications

Includes 1 qt. trigger spray bottle
Color Pink
Product Type Surface and Mat Cleaner
Material Plastic
For Use With (Application) Cleaning surfaces and mats
Certifications/Compliance RoHS Compliant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.