Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desco™ Reztore Surface and Mat Cleaner, 2.5 Gallon Refill
SDP

Catalog No. 18688045
Encompass
Encompass_Preferred
Change view
Click to view available options
Includes:
2.5 gal. refill box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
18-688-045 2.5 gal. refill box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-688-045 Supplier Desco™ Supplier No. 10448
May include imposed supplier surcharges.
Add to Cart
Add to Cart

ESD Surface and Mat Cleaner is used to clean and maintain ESD mats and other worksurfaces.

  • 2.5 Gallon Reztore™ Refill System
  • Refills 10446 and 10447 Reztore™ Alcohol-Free ESD Surface and Mat Cleaner spray bottles
  • Bag-in-box packaging
  • Lead-free RoHS compliant

Specifications

Includes 2.5 gal. refill box
Color Pink
Product Type Surface and Mat Cleaner
Material Cardboard
For Use With (Application) Cleaning surfaces and mats
Certifications/Compliance RoHS Compliant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.