Learn More
MOLECULAR DEPOT LLC Folic Acid BSA - 0.5 mg
Supplier:  MOLECULAR DEPOT LLC P2010026
Encompass_Preferred
Folic Acid BSA Conjugate Solution is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to Folic Acid (folate when in anionic form) Each BSA molecule contains several Folic Acid units Dilutions of the provided conjugate can be made in Phosphate Buffer Saline For in vitro research use only
Catalog No. 50-000-13352
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.