Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFC2 Antibody, DyLight 594, R&D Systems™
SDP

Catalog No. NB243535
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB243535 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB243535 Supplier R&D Systems Supplier No. NB100231DL594

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

RFC2 Polyclonal specifically detects RFC2 in Human samples. It is validated for Western Blot.

Specifications

Antigen RFC2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias A1, A1 40 kDa subunit, Activator 1 40 kDa subunit, Activator 1 subunit 2, MGC3665, replication factor C (activator 1) 2, 40kDa, replication factor C 2, Replication factor C 40 kDa subunit, replication factor C subunit 2, RF-C 40 kDa subunit, RFC40replication factor C (activator 1) 2 (40kD)
Host Species Goat
Immunogen The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Replication Factor C subunit 2 using the numbering given in entry NP_852136.1 (GeneID 5982).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5982
Target Species Human
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.